(±)-δ-Tetradecalactone ≥98%

Warning

B24794.14 B24794.22
B24794.14EA 34.7 EUR
B24794.14 B24794.22
(±)-δ-Tetradecalactone ≥98%
(±)-δ-Tetradecalactone

Purity: 98% CAS No.: 2721-22-4 Molecular Formula: C14H28O2 Formula Weight: 226.36 Flash Point: 110°(230°F) Melting Point: 130-135° MDL No.: MFCD00059809

Formula: C₁₄H₂₆O₂
Melting Pt: 130…135 °C
Density: 0,935
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059809
CAS Number: 2721-22-4
EINECS: 220-334-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR