Anti-IgG Rabbit Polyclonal Antibody (CF®583R)

Supplier: Biotium
20911-50UL 20911-500UL
BTIU20911-50ULEA 103 EUR
BTIU20911-50UL BTIU20911-500UL
Anti-IgG Rabbit Polyclonal Antibody (CF®583R)
Antibodies
This is a highly cross-adsorbed rabbit anti-mouse IgG (H L) secondary antibody labelled with our bright and photostable CF® Dyes. To minimize cross-reactivity, the antibody has been adsorbed against human serum.

  • Highly cross-adsorbed for specific staining with minimal background
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
  • CF® Dye and biotin conjugates: add 0,5 ml dH₂O

Liquid format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended. Liquid format antibodies contain 50% glycerol and will not freeze at −20 °C.

Lyophilised format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended.

Add the indicated volume of water directly to the vial containing the lyophilised antibody and mix gently to dissolve. Store reconstituted antibody at −20 °C and protect from light. Aliquot to avoid repeated freeze/thaw cycles. Alternatively, an equal volume of glycerol can be mixed with the reconstituted antibody so that it will remain liquid at −20 °C.

Type: Secondary
Antigen: IgG (Mouse)
Clonality: polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR