Anti-Donkey Polyclonal Antibody (CF® 583R)

Supplier: Biotium
20894-50UL 20894-500UL
BTIU20894-50ULEA 103 EUR
BTIU20894-50UL BTIU20894-500UL
Anti-Donkey Polyclonal Antibody (CF® 583R)
Antibodies
Highly cross-adsorbed donkey anti-mouse IgG (H L) secondary antibody labeled with our superior CF® dyes and other labels.

  • Highly cross-adsorbed for specific staining with minimal background
  • Available in 16 bright and photostable CF® dyes
  • Biotin, AP, and HRP conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology

This is a highly cross-adsorbed donkey anti-mouse IgG (H L) secondary antibody labeled with our bright and photostable CF® dyes and other labels. To minimise cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, syrian hamster, horse, human, rabbit, rat and sheep serum proteins.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse IgG
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR