2,4-Dimethylstyrene 97% stabilised
Supplier: ACROS ORGANICS
|
Warning
|
271150050
271150250
ACRO271150050EA
91.9
EUR
ACRO271150050
ACRO271150250
2,4-Dimethylstyrene 97% stabilised
2,4-Dimethylstyrene
Formula:
C₁₀H₁₂ Boiling Pt: 73…74 °C (10 mmHg) Melting Pt: –64 °C Density: 0,906 g/cm³ Flash Pt: 60 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014937 CAS Number: 2234-20-0 EINECS: 218-781-3 UN: 3295 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5410 to 1.5440 (20°C, 589 nm) |
GC | ≥96.0 % |
Stabilizer | ca. 0.05 % t-Butylcatechol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...