4-Methoxystyrene, Sigma-Aldrich®

Supplier: MERCK MILLIPORE


M-Clarity™ quality level = MQ200

8.41398.0005 8.41398.0025
8.41398.0005EA 144 EUR
8.41398.0005 8.41398.0025
4-Methoxystyrene, Sigma-Aldrich®
4-Methoxystyrene
Formula: C₉H₁₀O
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 3,3 °C
Density: 1,002 g/cm³ (20 °C)
MDL Number: MFCD00008619
CAS Number: 637-69-4
EINECS: 211-298-9

Order Now

Specification Test Results

Molar Mass 134.18 g/mol
Boiling Point 204 - 205 °C
Flash Point 76 °C
Density 1.002 g/cm3 (20 °C)
Synonyms 4-Vinylanisole
Hill Formula C9H10O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR