Ethylenetetracarbonitrile ≥98%
Supplier: ALFA AESAR
|
Danger
|
A13945.03
A13945.06
A13945.03EA
23.7
EUR
A13945.03
A13945.06
Ethylenetetracarbonitrile ≥98%
Ethylenetetracarbonitrile
Purity: 98%
CAS No.: 670-54-2
Molecular Formula: C6N4
Formula Weight: 128.09
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Density: 1.348
Sensitivity: Moisture Sensitive
Melting Point: 197-199°
Boiling Point: 223°
Beilstein Reference: 1679885
Merck Reference: 14,9195
UN No.: 3439
MDL No.: MFCD00001850
Formula:
C₆N₄ Boiling Pt: 223 °C Melting Pt: 197…199 °C Density: 1,348 |
MDL Number:
MFCD00001850 CAS Number: 670-54-2 EINECS: 211-578-0 UN: 3439 ADR: 6.1,II Merck Index: 14,9195 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...