Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (APC)
Supplier: STEMCELL TECHNOLOGIES
60011AZ 60011AZ.1
STMC60011AZEA 0 EUR
STMC60011AZ STMC60011AZ.1
Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (APC)
Antibodies
Mouse monoclonal IgG1 antibody against human, chimpanzee CD3, APC­-conjugated.
  • T cells
The UCHT1 antibody reacts with the ~20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ~95% of mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of TMore Product Information