Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (PE)

Supplier: STEMCELL TECHNOLOGIES
100-0311 100-0313
STMC100-0311EA 0 EUR
STMC100-0311 STMC100-0313
Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (PE)
Antibodies
Mouse monoclonal IgG2a, antibody against human, rhesus, cynomolgus HLA-DR, PE-conjugated.

  • B cells
  • Macrophages
  • Monocytes
  • T cells

The L243 antibody reacts with human HLA-DR, the major histocompatibility complex (MHC) class II receptor and a member of the immunoglobulin protein superfamily. HLA-DR is a heterodimeric transmembrane glycoprotein comprising a 36-kDa α subunit associated non-covalently with a 27-kDa β subunit and is expressed on the surface of antigen-presenting cells such as B cells, activated T cells, monocytes, macrophages and dendritic cells, activated natural killer (NK) cells, and progenitor cells. Together with the CD3/T cell receptor (TCR) complex and CD4 molecules, HLA-DR mediates a critical function in presenting peptides generated from hydrolysis of exogenous antigens by antigen-presenting cells to CD4+T (helper) cells, thereby either suppressing or inducing an immune response to the peptides. Thus, HLA-DR is involved in graft-versus-host disease and several autoimmune conditions. The L243 antibody binds an extracellular, conformational, and nonpolymorphic epitope on the α chain that is expressed only when the αβ heterodimer is correctly folded, and binding is not dependent on peptide loading of HLA-DR. Binding of the antibody blocks the association of HLA-DR with TCRs and reportedly exerts cytotoxic effects on human cells. The L243 antibody does not cross-react with HLA-DQ or HLA-DP.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-HLA-DR Mouse Monoclonal Antibody [clone: L243] (PE)
Antigen: HLA-DR
Clonality: Monoclonal
Clone: L243
Conjugation: PE
Epitope:
Host: Mouse
Isotype:
Reactivity: African green monkey, Baboon, Chimpanzee, Common marmoset, Cotton-topped tamarin, Cynomolgus monkey, Dog, Human, Pigtailed macaque, Rhesus monkey, Squirrel monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR