Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4] (FITC)

Supplier: STEMCELL TECHNOLOGIES
60096FI 60096FI.1
STMC60096FIEA 0 EUR
STMC60096FI STMC60096FI.1
Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4] (FITC)
Antibodies
Mouse monoclonal IgG1 antibody against human, chimpanzee CD33, FITC-conjugated.

  • Granulocytes
  • Subsets
  • Myeloid cells

The HIM3-4 antibody reacts with CD33 (Siglec-3), an ~67 - 75 kDa type 1 transmembrane sialoadhesion protein of the immunoglobulin superfamily. CD33 is expressed on committed myeloid progenitor cells, monocytes, granulocytes, dendritic cells, mast cells, resident macrophages, and subsets of B cells and activated T and NK cells. CD33 is a major marker used to differentiate myeloid from lymphoid or erythroid leukemias. In peripheral blood it is expressed at high levels on monocytes and at lower levels on granulocytes. CD33 functions as a sialic acid-dependent cell adhesion molecule and mediates protein-glycan and protein-protein interactions between cells. It also has a putative role as an inhibitory receptor during the immune response. HIM3-4 reportedly binds to the extracellular C2 Ig domain of CD33, and does not block subsequent binding of several other anti-CD33 clones, including P67.6, WM53, and WM54. Two isoforms designated CD33M and CD33m have been identified and among these clones only HIM3-4 recognizes both isoforms.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD33
Clonality: Monoclonal
Clone: HIM3-4
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR