3-Amino-2-naphthoic acid, Technical Grade
Supplier: ALFA AESAR
B23927.03
B23927.06
B23927.03EA
0
EUR
B23927.03
B23927.06
3-Amino-2-naphthoic acid, Technical Grade
3-Amino-2-naphthoic acid
Formula:
C₁₁H₉NO₂ Boiling Pt: 383 °C (1013 hPa) Melting Pt: 216…217 °C |
MDL Number:
MFCD00004115 CAS Number: 5959-52-4 EINECS: 227-726-2 Merck Index: 13,00451 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...