Furnaces, CWF series
Supplier: CARBOLITE
CWF115301 CWF1113301 CWF1123301 CWF125301 CWF1213301 CWF1223301 CWF135301 CWF1313301 CWF1323301 CWF1153216P1 CWF11133216P1 CWF11233216P1 CWF1253216P1 CWF12133216P1 CWF1353216P1 CWF13133216P1 CWF13233216P1 CWF12233508P1 CWF12233216P1 CWF11133508P1 CWF11233508P1 CWF1153508P1 CWF12133508P1 CWF1253508P1 CWF13133508P1 CWF13233508P1 CWF1353508P1 CWF1313-230SN CWF1323-230SN CWF1323-400TN CWF1123-230SN CWF1123-400TN CWF1105-230SN CWF1213-230SN CWF1223-230SN CWF1223-400TN CWF1205-230SN CWF1305-230SN
455-3121EA 3310 EUR
455-3121 455-3122 455-3123 CWF12/5/301 CWF12/13/301/ 455-3126 455-3127 455-3128 455-3129 455-0013 455-0009 455-0011 455-0019 455-0015 455-0025 455-0021 455-0023 CARBCWF12233508P1 CARBCWF12233216P1 CARBCWF11133508P1 CARBCWF11233508P1 CARBCWF1153508P1 CARBCWF12133508P1 CARBCWF1253508P1 CARBCWF13133508P1 CARBCWF13233508P1 CARBCWF1353508P1 CARBCWF1313-230SN CARBCWF1323-230SN CARBCWF1323-400TN CARBCWF1123-230SN CARBCWF1123-400TN CARBCWF1105-230SN CARBCWF1213-230SN CARBCWF1223-230SN CARBCWF1223-400TN CARBCWF1205-230SN CARBCWF1305-230SN
Furnaces, CWF series
Ovens Furnaces (Ashing and Muffle Ovens)
The CWF furnaces heating modules are situated on each side of the chamber and house a free radiating element, which offers 50% faster performance in comparison with traditional embedded element designs. Hard wearing materials are integral to the door surround and hearth, providing excellent resistance to everyday wear and tear, whilst secondary low thermal mass insulation ensures maximum thermal efficiency.
  • Safe operation ensured by double skin construction providing natural air convection for low outer case temperatures
  • Power to the elements is isolated
More Product Information