Furnaces economy, ELF series
Supplier: CARBOLITE
ELF1114B301 ELF11/6B ELF1123B ELF1106-230SN ELF1114-230SN ELF1123-230SN ELF1123-400TN
455-0026EA 2570 EUR
455-0026 ELF11/6B 455-0132 CARBELF1106-230SN CARBELF1114-230SN CARBELF1123-230SN CARBELF1123-400TN
Furnaces economy, ELF series
Ovens Furnaces (Ashing and Muffle Ovens)
The ELF furnaces are ideal for light heat treatment applications and for educational purposes, they are not suitable for ashing or heavy industrial duties; please see details on the AAF and CWF ranges respectively. Digital PID temperature controller (type 301) ensures precise and repeatable temperatures with push button temperature setting, adjustable heating rate and a process timer.
  • Safety features include; door switch that isolates power from the heating elements and a double skinned body for safe outer surface temperatures
  • Removable dense ceramic chamber hearth floor for easy spillage clean up
  • Drop down door may be used as a shelf when loading/unloading
These models have two side mounted free radiating elements embedded in low thermal mass insulation, which ensures rapid heat up, while providing excellent thermal uniformity within the chamber.
Delivery information: Note these furnaces are supplied without plugs and should be hardwired at position of use. The ELF 11/6 and ELF11/14 models require 230 V single phase and ELF 11/23 model requires 230 V single phase or can alternatively be supplied with 415 V two phase (of three phase) supply at extra charge.
Caution: Note: Maximum continuous operating temperature should be 100 °C below the max. temp. of the model shown above.