1,2,3-Tribromopropane ≥98%
|
Danger
|
L03752.14
L03752.22
L03752.14EA
21.3
EUR
L03752.14
L03752.22
1,2,3-Tribromopropane ≥98%
1,2,3-Tribromopropane
Purity: 98%
CAS No.: 96-11-7
Molecular Formula: C3H5Br3
Formula Weight: 280.80
Flash Point: 93°(199°F)
Refractive Index: 1.5850
Melting Point: 16-17°
Boiling Point: 220°
Beilstein Reference: 1732082
Merck Reference: 14,9614
MDL No.: MFCD00017884
Formula:
BrH₂CBrCHCH₂Br MW: 280.78 g/mol Boiling Pt: 220 °C Melting Pt: 16…17 °C Density: 2.398 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017884 CAS Number: 96-11-7 EINECS: 202-478-8 Merck Index: 12,09745 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...