1-Bromo-5-chloropentane 98%
Supplier: ACROS ORGANICS
|
Warning
|
221210050
221210250
221211000
ACRO221210050EA
28.5
EUR
ACRO221210050
ACRO221210250
ACRO221211000
1-Bromo-5-chloropentane 98%
1-Bromo-5-chloropentane
Formula:
BrH₂C(CH₂)₃CH₂Cl MW: 185.49 g/mol Boiling Pt: 210…212 °C (760 mmHg) Density: 1.4 g/cm³ Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001016 CAS Number: 54512-75-3 EINECS: 259-193-7 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4826 to 1.4846 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...