Cleanroom gloves, latex, CERTICLEAN®
Supplier: KIMBERLY CLARK
40101-160_INT 40101-162_INT 40101-164_INT
KIMB40101-160_INTEA 0 EUR
KIMB40101-160_INT KIMB40101-162_INT KIMB40101-164_INT
Cleanroom gloves, latex, CERTICLEAN®
Gloves
Ambidextrous gloves with beaded cuffs for strength and reduction of cuff roll down.
  • Non sterile, powder-free
  • Film thickness designed for comfort and tactile sensitivity
  • Textured for a reliable grip
  • Silicone-free material minimize ionic contaminants and non volatile residue (NVR) transfer
Suitable for use in Class 100, ISO 5, M3.5 cleanrooms.
Packaging: Packaged in cleanroom-compatible polyethylene, 100 gloves/double poly bag, 10 bags/case.