Ethylenediamine monohydrate, Sigma-Aldrich®
|
Danger
|
M-Clarity™ quality level = MQ200
8.20030.0100
8.20030.1000
8.20030.0100EA
47.1
EUR
8.20030.0100
8.20030.1000
Ethylenediamine monohydrate, Sigma-Aldrich®
Ethylenediamine monohydrate
Formula:
C₂H₈N₂·1H₂O Boiling Pt: 118 °C (1013 hPa) Melting Pt: 11 °C Density: 0,926 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149563 CAS Number: 6780-13-8 EINECS: 203-468-6 UN: 1604 ADR: 8 (3),II Merck Index: 13,03829 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 8 - 10 °C |
Molar Mass | 78.11 g/mol |
Boiling Point | 118 °C (1013 hPa) |
Vapor Pressure | 12 hPa (20 °C) |
Flash Point | 42 °C |
Explosion Limit | 2.7 - 16.6 %(V) |
Density | 0.963 g/cm3 (20 °C) |
pH | 12 (100 g/l, H2O, 20 °C) |
Ignition Point | 385 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...