Sodium cyclamate ≥98%

Supplier: ALFA AESAR
Warning

A18666.0B A18666.0I A18666.30
A18666.0BEA 137 EUR
A18666.0B A18666.0I A18666.30
Sodium cyclamate ≥98%
Sodium cyclamate

Purity: 98% CAS No.: 139-05-9 Molecular Formula: C6H12NNaO3S Formula Weight: 201.22 Melting Point: 300° Beilstein Reference: 4166868

Formula: C₆H₁₂NNaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003827
CAS Number: 139-05-9
EINECS: 205-348-9
Merck Index: 12,02770

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR