Methoxyacetic acid ≥97%

Supplier: ALFA AESAR
Danger

B22917.0B B22917.18 B22917.30
B22917.0BEA 230 EUR
B22917.0B B22917.18 B22917.30
Methoxyacetic acid ≥97%
Methoxyacetic acid

Purity: 97% CAS No.: 625-45-6 Molecular Formula: C3H6O3 Formula Weight: 90.08 Flash Point: 114°(237°F) Refractive Index: 1.4165 Odour: Odourless pH: 1.6 Melting Point: 8° Boiling Point: 202-204° Beilstein Reference: 635800 UN No.: 3265 MDL No.: MFCD00004308

Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 202…204 °C
Melting Pt: 8 °C
Density: 1.174
Flash Pt: 114 °C (237 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004308
CAS Number: 625-45-6
EINECS: 210-894-6
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR