Chloroform ≥99% stabilised
|
Danger
|
Stabilization:
Stabilised with ethanol 1 %
L13200.AP
L13200.AU
L13200.APEA
30.1
EUR
L13200.AP
L13200.AU
Chloroform ≥99% stabilised
Chloroform
Stabilized with 0.8-1% ethanol
Purity: 99%
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Refractive Index: 1.4440
Odour: Characteristic
Boiling Point: 60.5-61.5°
Beilstein Reference: 1731042
Merck Reference: 14,2141
UN No.: 1888
MDL No.: MFCD00000826
Formula:
CHCl₃ Boiling Pt: 60,5…61.5 °C Melting Pt: –63 °C Density: 1,492 Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1,III REACH: 05-2114614906-45 Merck Index: 14,2141 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...