2,2,3,3-Tetrafluoro-1-propanol ≥97%

Warning

B21594.09 B21594.18 B21594.30
B21594.09EA 39.6 EUR
B21594.09 B21594.18 B21594.30
2,2,3,3-Tetrafluoro-1-propanol ≥97%
2,2,3,3-Tetrafluoro-1-propanol

Purity: 97% CAS No.: 76-37-9 Molecular Formula: H(CF2)2CH2OH Formula Weight: 132.06 Flash Point: 49°(120°F) Refractive Index: 1.3195 Melting Point: -70° Boiling Point: 107-109° Beilstein Reference: 1738264 Merck Reference: 14,9206 UN No.: 1987 MDL No.: MFCD00004676

Formula: C₃H₄F₄O
Boiling Pt: 107…109 °C
Melting Pt: < –70 °C
Density: 1,471
Flash Pt: 49 °C (120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004676
CAS Number: 76-37-9
EINECS: 200-955-5
UN: 1987
ADR: 3,III
Merck Index: 14,9206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR