1-Iododecane ≥98% stabilised

Supplier: ALFA AESAR
Warning


Stabilization: Stabilised with copper

A16173.14 A16173.18 A16173.30
A16173.14EA 42.2 EUR
A16173.14 A16173.18 A16173.30
1-Iododecane ≥98% stabilised
1-Iododecane

Stabilized with copper


Purity: 98% CAS No.: 2050-77-3 Molecular Formula: C10H21I Formula Weight: 268.18 Flash Point: 96°(204°F) Refractive Index: 1.4850 Melting Point: -16° Boiling Point: 264° Beilstein Reference: 1735228 MDL No.: MFCD00001085

Formula: C₁₀H₂₁I
Boiling Pt: 264 °C
Melting Pt: –16 °C
Density: 1,257
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001085
CAS Number: 2050-77-3
EINECS: 218-104-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR