Sodium nonyl sulphate ≥99%

Warning

L06418.03 L06418.06
L06418.03EA 51.7 EUR
L06418.03 L06418.06
Sodium nonyl sulphate ≥99%
Sodium nonyl sulphate

Purity: 99% CAS No.: 1072-15-7 Molecular Formula: CH3(CH2)8OSO3Na Formula Weight: 246.30 Beilstein Reference: 3748751 MDL No.: MFCD00024893

Formula: C₉H₁₉NaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00024893
CAS Number: 1072-15-7
EINECS: 214-002-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR