n-Pentane ≥98% Environmental grade
Supplier: ALFA AESAR
|
Danger
|
40981.K2
40981.K7
40981.M6
40981.K2EA
0
EUR
40981.K2
40981.K7
40981.M6
n-Pentane ≥98% Environmental grade
n-Pentane
Purity: 98+%
CAS No.: 109-66-0
Molecular Formula: CH3(CH2)3CH3
Formula Weight: 72.15
Flash Point: -49°(-57°F)
Physical Form: Liquid
Density: 0.626
Refractive Index: 1.3580
Appearance: Colourless
Melting Point: -130°
Boiling Point: 36°
Merck Reference: 14,7116
UN No.: 1265
MDL No.: MFCD00009498
Suitable for environmental, pesticide, and GC analysis
Formula:
H₃C(CH₂)₃CH₃ MW: 72.15 g/mol Boiling Pt: 36 °C Melting Pt: –130 °C Density: 0.626 Flash Pt: –49 °C (–57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3 II REACH: 01-2119459286-30 Merck Index: 13,07193 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...