1-Tetradecene, Supelco®

Supplier: SUPELCO


M-Clarity™ quality level = MQ100

87187-25ML 87187-5ML
87187-25MLEA 238 EUR
87187-25ML 87187-5ML
1-Tetradecene, Supelco®
1-Tetradecene
Formula: C₁₄H₂₈
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: –13 °C
Density: 0,7708 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008981
CAS Number: 1120-36-1
EINECS: 214-306-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=200a2e4c0244030e459c4c087bfc6a0ef7d54fea&vid=200a2e4c0244030e459c4c087bfc6a0ef7d54fea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR