Ethyl-3,4-dimethoxybenzoate ≥98%
L02355.14
L02355.22
L02355.14EA
67.4
EUR
L02355.14
L02355.22
Ethyl-3,4-dimethoxybenzoate ≥98%
Ethyl 3,4-dimethoxybenzoate
Purity: 98+%
CAS No.: 3943-77-9
Molecular Formula: C11H14O4
Formula Weight: 210.23
Melting Point: 42-45°
Beilstein Reference: 3118897
Formula:
C₁₁H₁₄O₄ Melting Pt: 42…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015150 CAS Number: 3943-77-9 EINECS: 223-526-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...