3-Bromophenol 98%
Supplier: ACROS ORGANICS
|
Warning
|
146340100
146340250
146341000
ACRO146340100EA
39.7
EUR
ACRO146340100
ACRO146340250
ACRO146341000
3-Bromophenol 98%
3-Bromophenol
Formula:
C₆H₅BrO Boiling Pt: 236 °C (760 mmHg) Melting Pt: 30…32 °C Density: 0,641 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002253 CAS Number: 591-20-8 EINECS: 209-706-5 ADR: 6.1,III Merck Index: 13,01413 |
Specification Test Results
Appearance | Clear colorless to yellow to brown Liquid after melting or solid |
Infrared spectrum | Conforms |
Refractive index | 1.5950 to 1.6010 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...