2,4,6-Trifluorobenzonitrile ≥99%
Supplier: ALFA AESAR
|
Danger
|
L15525.06
L15525.03
L15525.06EA
0
EUR
L15525.06
L15525.03
2,4,6-Trifluorobenzonitrile ≥99%
2,4,6-Trifluorobenzonitrile
Purity: 99%
CAS No.: 96606-37-0
Molecular Formula: C7H2F3N
Formula Weight: 157.10
Flash Point: 92°(197°F)
Melting Point: 55-58°
Boiling Point: 179°
Beilstein Reference: 5512504
UN No.: 3439
MDL No.: MFCD00042399
Formula:
C₇H₂F₃N Boiling Pt: 179 °C Melting Pt: 55…58 °C Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042399 CAS Number: 96606-37-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...