4-Fluoroanthranilic acid 96%

Supplier: ACROS ORGANICS
Warning

293020025 293020100
ACRO293020025EA 122 EUR
ACRO293020025 ACRO293020100
4-Fluoroanthranilic acid 96%
4-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 188…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00075553
CAS Number: 446-32-2
EINECS: 207-163-9

Order Now

Specification Test Results

Appearance White to beige to light brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 187 to 195 °C
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR