1-Boc-4-piperidone ≥99%
L13361.09
L13361.18
L13361.09EA
82.9
EUR
L13361.09
L13361.18
1-Boc-4-piperidone ≥99%
1-Boc-4-piperidone
Purity: 99%
CAS No.: 79099-07-3
Molecular Formula: C10H17NO3
Formula Weight: 199.25
Melting Point: 72-76°
Beilstein Reference: 3650236
Formula:
C₁₀H₁₇NO₃ Melting Pt: 72…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151800 CAS Number: 79099-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...