Thiophenol, Supelco®

Danger


M-Clarity™ quality level = MQ100

09286-25ML-F 09286-5ML-F
09286-25ML-FEA 249 EUR
09286-25ML-F 09286-5ML-F
Thiophenol, Supelco®
Thiophenol
Formula: C₆H₆S
MW: 110.18 g/mol
Boiling Pt: 169
Melting Pt: –15 °C
Density: 1.0728 g/cm³ (20 °C)
Flash Pt: 50
Storage Temperature: Ambient
MDL Number: MFCD00004826
CAS Number: 108-98-5
EINECS: 203-635-3
UN: 2337
ADR: 6.1 I
Merck Index: 12,09492
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d36ebc3ab9acd950e88ec80fbf2956ef8019bd49&vid=d36ebc3ab9acd950e88ec80fbf2956ef8019bd49&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR