Cumene, Supelco®

Supplier: SUPELCO
Danger


M-Clarity™ quality level = MQ100

28220-5ML 28220-25ML
28220-5MLEA 59.7 EUR
28220-5ML 28220-25ML
Cumene, Supelco®
Cumene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 151
Melting Pt: –96 °C
Density: 0.8624 g/cm³ (20 °C)
Flash Pt: 31
Storage Temperature: Ambient
MDL Number: MFCD00008881
CAS Number: 98-82-8
EINECS: 202-704-5
UN: 1918
ADR: 3 III
Merck Index: 13,02644
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a579b2508c688582eda1774b2166699c4e1773c&vid=5a579b2508c688582eda1774b2166699c4e1773c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR