Bis(triphenylphosphine)nickel(II) chloride ≥98%
|
Danger
|
13930.09
13930.18
13930.09EA
60.6
EUR
13930.09
13930.18
Bis(triphenylphosphine)nickel(II) chloride ≥98%
Bis(triphenylphosphine)nickel (II) chloride
Purity: 98%
CAS No.: 14264-16-5
Molecular Formula: NiCl2[P(C6H5)3]2
Formula Weight: 654.18
Physical Form: Crystalline powder
Sensitivity: Moisture Sensitive
Appearance: Grey to green to black
Odour: Odourless
Melting Point: ca 225° dec.
MDL No.: MFCD00009592
Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron
Formula:
C₃₆H₃₂Cl₂NiP₂ MW: 656.19 g/mol Melting Pt: ∼ 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00009592 CAS Number: 14264-16-5 EINECS: 238-154-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...