(±)-Methyl-2-bromobutyrate ≥97%
Supplier: ALFA AESAR
A19468.0B
A19468.30
A19468.0BEA
0
EUR
A19468.0B
A19468.30
(±)-Methyl-2-bromobutyrate ≥97%
(±)-Methyl 2-bromobutyrate
Purity: 97%
CAS No.: 3196-15-4
Molecular Formula: C5H9BrO2
Formula Weight: 181.04
Flash Point: 68°(154°F)
Refractive Index: 1.4520
Boiling Point: 137-138°/50mm
Beilstein Reference: 1720826
UN No.: 1760
MDL No.: MFCD00009666
Formula:
C₅H₉BrO₂ Flash Pt: 68 °C (154 °F) |
MDL Number:
MFCD00009666 CAS Number: 3196-15-4 EINECS: 221-699-0 ADR: , |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...