Bismuth(III) chloride, ultra dry ≥99.997% (metals basis)
|
Danger
|
Ampuled under argon
35680.06
35680.14
35680.06EA
116
EUR
35680.06
35680.14
Bismuth(III) chloride, ultra dry ≥99.997% (metals basis)
Bismuth (III) chloride
Purity: 99.997+%
CAS No.: 7787-60-2
Molecular Formula: BiCl3
Formula Weight: 315.34
Physical Form: Powder, Ampouled under argon
Density: 4.75
Appearance: Off-white
Melting Point: 230-232°
Boiling Point: ca 430° subl.
Merck Reference: 14,1262
UN No.: 3260
MDL No.: MFCD00003461
Formula:
BiCl₃ MW: 315.34 g/mol Boiling Pt: ∼430 °C (subl.) Melting Pt: 230…232 °C Density: 4.75 Storage Temperature: Ambient |
MDL Number:
MFCD00003461 CAS Number: 7787-60-2 EINECS: 232-123-2 UN: 3260 ADR: 8 III Merck Index: 14,1262 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...