Chloropentacarbonylrhenium 98%

Supplier: ACROS ORGANICS
Warning

316310010 316310050
ACRO316310010EA 215 EUR
ACRO316310010 ACRO316310050
Chloropentacarbonylrhenium 98%
Chloropentacarbonylrhenium
Formula: C₅ClO₅Re
MW: 361.71 g/mol
MDL Number: MFCD00013296
CAS Number: 14099-01-5
EINECS: 237-948-1
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to off-white Powder
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR