3-Cyclopentylpropionyl chloride ≥98%
Supplier: ALFA AESAR
|
Danger
|
A17715.14
A17715.22
A17715.14EA
73.5
EUR
A17715.14
A17715.22
3-Cyclopentylpropionyl chloride ≥98%
3-Cyclopentylpropionyl chloride
Purity: 98%
CAS No.: 104-97-2
Molecular Formula: C8H13ClO
Formula Weight: 160.64
Flash Point: 84°(184°F)
Refractive Index: 1.4645
Odour: Acrid
Boiling Point: 199-200°
Beilstein Reference: 1856952
UN No.: 3265
MDL No.: MFCD00001391
Formula:
C₈H₁₃ClO Boiling Pt: 199…200 °C Density: 1,049 Flash Pt: 84 °C (184 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001391 CAS Number: 104-97-2 EINECS: 203-257-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...