Phenylbutazone ≥98%
|
Danger
|
L03449.14
L03449.22
L03449.14EA
43.5
EUR
L03449.14
L03449.22
Phenylbutazone ≥98%
Phenylbutazone
Purity: 98%
CAS No.: 50-33-9
Molecular Formula: C19H20N2O2
Formula Weight: 308.38
Odour: Odourless
Melting Point: 105-108°
Beilstein Reference: 290080
Merck Reference: 14,7277
UN No.: 2811
MDL No.: MFCD00005500
Formula:
C₁₉H₂₀N₂O₂ Melting Pt: 105…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005500 CAS Number: 50-33-9 EINECS: 200-029-0 UN: 2811 ADR: 6.1,III Merck Index: 14,7277 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...