Methyl ethyl ketone, extra dry 99.5%, AcroSeal®

Supplier: ACROS ORGANICS
Danger

396951000 396950010
396951000.EA 98.4 EUR
396951000. 396950010.
Methyl ethyl ketone, extra dry 99.5%, AcroSeal®
Methyl ethyl ketone
Formula: C₂H₅COCH₃
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –87 °C
Density: 0,806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=7560bf484cae1b3d9ae7f8cc59b75018d61b5022&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR