4-Isopropylphenylacetonitrile ≥98%
Supplier: ALFA AESAR
|
Danger
|
L10715.03
L10715.06
L10715.03EA
38.2
EUR
L10715.03
L10715.06
4-Isopropylphenylacetonitrile ≥98%
4-Isopropylphenylacetonitrile
Purity: 98+%
CAS No.: 4395-87-3
Molecular Formula: C11H13N
Formula Weight: 159.23
Refractive Index: 1.5120
Boiling Point: 150°/25mm
UN No.: 3276
MDL No.: MFCD00040892
Formula:
C₁₁H₁₃N Boiling Pt: 150 °C (25 mmHg) Density: 0,960 Storage Temperature: Ambient |
MDL Number:
MFCD00040892 CAS Number: 4395-87-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...