AMPSO sodium salt

Supplier: ROTH CARL

7014.1 7014.2
ROTH7014.1EA 0 EUR
ROTH7014.1 ROTH7014.2
AMPSO sodium salt
AMPSO sodium salt
Formula: C₇H₁₆NNaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00067500
CAS Number: 102029-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR