N-Acetyl-β-D-glucosamine ≥98%
A13047.09
A13047.18
A13047.22
A13047.09EA
31.5
EUR
A13047.09
A13047.18
A13047.22
N-Acetyl-β-D-glucosamine ≥98%
N-Acetyl-β-D-glucosamine
Purity: 98+%
CAS No.: 7512-17-6
Molecular Formula: C8H15NO6
Formula Weight: 221.21
Storage Temperature: 2°C to 8°C
Melting Point: 213° dec.
Beilstein Reference: 1727589
Merck Reference: 14,4458
Optical Rotation: +42° (c=2 in water)
MDL No.: MFCD00136044
Formula:
C₈H₁₅NO₆ Melting Pt: 213 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00136044 CAS Number: 7512-17-6 EINECS: 231-368-2 Merck Index: 14,4458 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...