4'-(Trifluoromethyl)biphenyl-3-carboxylic acid ≥97%

Supplier: ALFA AESAR
Warning

H57246.MD H57246.03
H57246.MDEA 0 EUR
H57246.MD H57246.03
4'-(Trifluoromethyl)biphenyl-3-carboxylic acid ≥97%
4'-(Trifluoromethyl)biphenyl-3-carboxylic acid

Purity: 97% CAS No.: 199528-28-4 Molecular Formula: C14H9F3O2 Formula Weight: 266.22 MDL No.: MFCD03424604

Formula: C₁₄H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03424604
CAS Number: 199528-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR