Laboratory ovens, AX
Supplier: CARBOLITE
AX30 AX60 AX120 AX030-230SN AX060-230SN AX120-230SN
CARBAX30EA 1830 EUR
CARBAX30 CARBAX60 CARBAX120 CARBAX030-230SN CARBAX060-230SN CARBAX120-230SN
Laboratory ovens, AX
Ovens Standard Ovens
The Apex AX range of 250 °C laboratory ovens comprises three bench-top sized models equipped with the R38 digital PID temperature controller.
  • Fan convection for rapid heating and excellent uniformity
  • Chemically resistant stainless steel liner
  • Two adjustable nickel-chrome plated wire shelves
  • Lever latch door and airtight silicone seal
With over-temperature protection, digital countdown timer to switch oven off and lockable door.
Certifications: Compliant with safety standard BS EN 61010-2-010:2003.