Industrial gloves, half-finger, ActivArmr® 07-111
Supplier: ANSELL HEALTH CARE
07-111/10 07-111/8
112-4908EA 0 EUR
112-4908 112-4909
Industrial gloves, half-finger, ActivArmr® 07-111
Gloves
The VibraGuard® gloves feature a nitrile impregnated coating on a cotton interlock/Gelfôm™ liner. They provide resistance to cuts, punctures and abrasion, as well as to grease and oil.
  • Soft gel foam padding in the thumb, index finger and palm area enhances the shock impact protection
  • No need to remove the glove to screw or handle very small parts like nails
  • Half-finger design facilitates pressing the trigger of power tools
Ideal for hand and pedestal grinders, vibrating poker, road drills, and applications including chipping hammers, heavy-dutyMore Product Information