5-Chlorovaleryl chloride ≥98%
Supplier: ALFA AESAR
|
Danger
|
H56237.22
H56237.18
H56237.22EA
448
EUR
H56237.22
H56237.18
5-Chlorovaleryl chloride ≥98%
5-Chlorovaleryl chloride
Purity: 98%
CAS No.: 1575-61-7
Molecular Formula: C5H8Cl2O
Formula Weight: 155.02
Flash Point: 91°(196°F)
Refractive Index: 1.464
Boiling Point: 38-39°
Beilstein Reference: 1745182
UN No.: 3265
MDL No.: MFCD00000758
Formula:
C₅H₈Cl₂O MW: 155.02 g/mol Boiling Pt: 38…39 °C Density: 1.2 Flash Pt: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000758 CAS Number: 1575-61-7 EINECS: 216-403-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...