4-Chlorobenzylideneacetone ≥98%
Supplier: ALFA AESAR
L05321.06
L05321.14
L05321.06EA
54
EUR
L05321.06
L05321.14
4-Chlorobenzylideneacetone ≥98%
4-Chlorobenzylideneacetone
Purity: 98%
CAS No.: 3160-40-5
Molecular Formula: C10H9ClO
Formula Weight: 180.64
Melting Point: 55-60°
Boiling Point: 145-147°/5mm
MDL No.: MFCD00018790
Formula:
C₁₀H₉ClO Boiling Pt: 145…147 °C (5 mmHg) Melting Pt: 55…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018790 CAS Number: 3160-40-5 EINECS: 221-610-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...