N-(5-Hexynyl)phthalimide ≥97%
Supplier: ALFA AESAR
H53489.06
H53489.14
H53489.06EA
94.9
EUR
H53489.06
H53489.14
N-(5-Hexynyl)phthalimide ≥97%
N-(5-Hexynyl)phthalimide
Purity: 97%
CAS No.: 6097-08-1
Molecular Formula: C14H13NO2
Formula Weight: 227.26
Melting Point: 68-72°
MDL No.: MFCD00671372
Formula:
C₁₄H₁₃NO₂ Melting Pt: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671372 CAS Number: 6097-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...