4'-Isopropylacetanilide ≥97%
B21591.06
B21591.14
B21591.06EA
46
EUR
B21591.06
B21591.14
4'-Isopropylacetanilide ≥97%
4'-Isopropylacetanilide
Purity: 97+%
CAS No.: 5702-74-9
Molecular Formula: C11H15NO
Formula Weight: 177.25
Melting Point: 103-105°
MDL No.: MFCD00026406
Formula:
C₁₁H₁₅NO |
MDL Number:
MFCD00026406 CAS Number: 5702-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...