Sapphire Pipette Tips and Filter Tips
Supplier: GREINER BIO ONE
774353_5760 773353_5760 773363_5760 771350_10000 771355_4800 771351_5760 771352_5760
GREI774353_5760EA 1120 EUR
GREI774353_5760 GREI773353_5760 GREI773363_5760 GREI771350_10000 GREI771355_4800 GREI771351_5760 GREI771352_5760
Sapphire Pipette Tips and Filter Tips
Pipette Tips
Flexibility is critical in terms of modern liquid handling solutions. This is why the Sapphire tips are manufactured in eight different sizes, including an extended length 10 µl tip for the recovery of small sample volumes.
  • Fully automated production and packaging
  • Universal fit for all common pipettes
  • Tips (without filter) and racks are autoclavable
  • Filters manufactured without additives
  • Easy-to-read graduations
  • Coloured box inserts for easy volume identification
  • Low retention surface properties for high recovery rate and maximum precision
All our tips are made of medical grade polypropylene and are available as standard, low-retention, filter, or low-retention filter tips.
All Sapphire tips meet the same high-quality, uniform standards regardless of type, surface, or shape. Greiner Bio-One certifies that all tips are manufactured and tested in accordance with international standards and meet the required specifications. They are free of detectable DNase, RNase, human DNA, PCR inhibitors, and endotoxins (pyrogens). Additionally, they are non-cytotoxic and tested according to ISO 8655-6.
Depending on the format, Sapphire pipette tips are packed in racks, bulk, or refill units. When used together, Sapphire tips and pipettes provide a fully optimised and harmonised liquid handling solution.