Single Zone tube furnaces, MTF wire wound
Supplier: CARBOLITE
M1015130230SN M1025130230SN M1225250230SN M1225400230SN M1238250230SN M1238400230SN
CARBM1015130230SNEA 0 EUR
CARBM1015130230SN CARBM1025130230SN CARBM1225250230SN CARBM1225400230SN CARBM1238250230SN CARBM1238400230SN
Single Zone tube furnaces, MTF wire wound
Ovens Tube Furnaces
Tube furnaces are used to heat a small sample that features insulation plugs, radiation shields to prevent heat loss and improve uniformity (recommended for vertical use) and impervious inner worktubes to protect against chemical attack or damage from thermal shock.
  • Control module, with furnace mounted directly on top
  • Carbolite 301 controller, with single ramp to set-point facility
  • Horizontal tube configuration
  • Non-permeable inner wirewound worktube to contain modified atmosphere
  • Over-temperature protection
  • Delayed start / process timer function as standard
Rapid temperature changes are possible by simply using a push-rod to move the sample along the length of the tube (although care must be taken not to cause thermal shock to the tube or sample boat).